accessibility menu, dialog, popup

UserName

H3COH

CAS RN 70568-60-4

IUPAC Name: 4-butylcyclohexan-1-ol
Synonyms: 4-butylcyclohexan-1-ol
Molecular Weight (g/mol): 156.27
Molecular Formula: C10H20O
InChi Key: NUANGSLQWFBVEH-UHFFFAOYSA-N
SMILES: CCCCC1CCC(O)CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results