accessibility menu, dialog, popup

UserName

CAS RN 70289-12-2

CH3OHOO

CAS RN 70289-12-2

IUPAC Name: 2-(2-ethoxyphenyl)acetic acid
Synonyms: (2-ethoxyphenyl)acetic acid
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: ABROWERIMKBDEW-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC=C1CC(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results