missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyphenylacetic Acid 98.0+%, TCI America™

Catalog No. E12345G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12345G 5 g
E12341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E12345G Supplier TCI America Supplier No. E12345G
Only null left

Chemical Identifiers

CAS 70289-12-2
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00016824
InChI Key ABROWERIMKBDEW-UHFFFAOYSA-N
PubChem CID 144397
IUPAC Name 2-(2-ethoxyphenyl)acetic acid
SMILES CCOC1=CC=CC=C1CC(O)=O

Specifications

Melting Point 106°C
Color White
Quantity 5 g
Formula Weight 180.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Ethoxyphenylacetic Acid