accessibility menu, dialog, popup

UserName

NBrOBr

CAS RN 70201-42-2

IUPAC Name: 3,5-dibromopyridine-4-carbaldehyde
Synonyms: 3,5-dibromopyridine-4-carbaldehyde
Molecular Weight (g/mol): 264.9
Molecular Formula: C6H3Br2NO
InChi Key: WPBYVMDYYFWYAY-UHFFFAOYSA-N
SMILES: BrC1=CN=CC(Br)=C1C=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results