accessibility menu, dialog, popup

UserName

CAS RN 7011-83-8

H3CH3COO

CAS RN 7011-83-8

IUPAC Name: 5-hexyl-5-methyloxolan-2-one
Synonyms: gamma-methyldecalactone
Molecular Weight (g/mol): 184.28
Molecular Formula: C11H20O2
InChi Key: ALWUKGXLBSQSMA-UHFFFAOYNA-N
SMILES: CCCCCCC1(C)CCC(=O)O1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results