accessibility menu, dialog, popup

UserName

NH2H2NOOHO

CAS RN 70-47-3

IUPAC Name: 2-amino-3-carbamoylpropanoic acid
Synonyms: asparagin asparagina Hasp
Molecular Weight (g/mol): 132.12
Molecular Formula: C4H8N2O3
InChi Key: DCXYFEDJOCDNAF-UHFFFAOYNA-N
SMILES: NC(CC(N)=O)C(O)=O
Percent Purity 
  • (1)
  • (7)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (6)
  • (1)
  • (7)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)