accessibility menu, dialog, popup

UserName

CAS RN 698-67-9

IUPAC Name: 4-bromobenzamide
Synonyms: 4-bromobenzamide
Molecular Weight (g/mol): 200.04
Molecular Formula: C7H6BrNO
InChi Key: ZRWNRAJCPNLYAK-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=C(Br)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)