missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzamide, 97%

Catalog No. AAB2434818
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2434818 50 g
AAB2434804 2 g
AAB2434809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2434818 Supplier Thermo Scientific Chemicals Supplier No. B2434818
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 698-67-9 | C7H6BrNO | 200.035 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 698-67-9
Molecular Formula C7H6BrNO
Molecular Weight (g/mol) 200.035
MDL Number MFCD00007991
InChI Key ZRWNRAJCPNLYAK-UHFFFAOYSA-N
Synonym p-bromobenzamide, benzamide, 4-bromo, benzamide, p-bromo, p-bromobenzoic acid amide, 4-bromo-benzamide, 4-bromo benzamide, pubchem3737, acmc-209obe, 4-bromobenzamide, 4-bromobenzoic acid amide
PubChem CID 69683
IUPAC Name 4-bromobenzamide
SMILES C1=CC(=CC=C1C(=O)N)Br

Specifications

Melting Point 190°C to 193°C
Quantity 50 g
Formula Weight 200.03
Percent Purity 97%
Chemical Name or Material 4-Bromobenzamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-817-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only