missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzamide 98.0+%, TCI America™

Catalog No. B42165G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42165G 5 g
B421625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42165G Supplier TCI America Supplier No. B42165G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 698-67-9
Molecular Formula C7H6BrNO
Molecular Weight (g/mol) 200.035
MDL Number MFCD00007991
InChI Key ZRWNRAJCPNLYAK-UHFFFAOYSA-N
Synonym p-bromobenzamide, benzamide, 4-bromo, benzamide, p-bromo, p-bromobenzoic acid amide, 4-bromo-benzamide, 4-bromo benzamide, pubchem3737, acmc-209obe, 4-bromobenzamide, 4-bromobenzoic acid amide
PubChem CID 69683
IUPAC Name 4-bromobenzamide
SMILES C1=CC(=CC=C1C(=O)N)Br

Specifications

Melting Point 193°C
Color White-Yellow
Quantity 5 g
Formula Weight 200.04
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromobenzamide
RTECSNumber CV2365800
TSCA No