accessibility menu, dialog, popup

UserName

CAS RN 6973-09-7

IUPAC Name: 5-amino-2-methylbenzene-1-sulfonamide
Synonyms: 5-amino-2-methylbenzenesulfonamide
Molecular Weight (g/mol): 186.23
Molecular Formula: C7H10N2O2S
InChi Key: KTPBKMYOIFHJMI-UHFFFAOYSA-N
SMILES: CC1=CC=C(N)C=C1S(N)(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)