missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-methylbenzenesulfonamide, 96%

Catalog No. AAH3405606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3405606 5 g
AAH3405603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3405606 Supplier Thermo Scientific Chemicals Supplier No. H3405606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6973-09-7 | C7H10N2O2S | 186.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6973-09-7
MDL Number MFCD06681071
ChEBI CHEBI:29013

Specifications

Melting Point 163°C to 164°C
Molecular Formula C31H48O6
Quantity 5 g
Synonym fusidic acid, fusidine, ramycin, fucithalmic, fucidic acid, fucidin acid, fusidate, fucidin, flucidin, fucidate
InChI Key IECPWNUMDGFDKC-MZJAQBGESA-N
SMILES CC1C2CCC3(C(C2(CCC1O)C)C(CC4C3(CC(C4=C(CCC=C(C)C)C(=O)O)OC(=O)C)C)O)C
IUPAC Name (2Z)-2-[(3R,4S,5S,8S,9S,10S,11R,13R,14S,16S)-16-acetyloxy-3,11-dihydroxy-4,8,10,14-tetramethyl-2,3,4,5,6,7,9,11,12,13,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-17-ylidene]-6-methylhept-5-enoic acid
Molecular Weight (g/mol) 516.72
PubChem CID 3000226
Formula Weight 186.23
Percent Purity 96%
Chemical Name or Material 5-Amino-2-methylbenzenesulfonamide
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only