CAS RN 697-90-5
IUPAC Name:
2,4-dichloro-6-iodoaniline
Synonyms:
2,4-dichloro-6-iodoaniline
Molecular Weight (g/mol):
287.91
Molecular Formula:
C6H4Cl2IN
InChi Key:
YJRSMJTVXWBFJJ-UHFFFAOYSA-N
SMILES:
NC1=C(Cl)C=C(Cl)C=C1I
1
–
1
of
1
results
1
Promotions Available
2,4-Dichloro-6-iodoaniline, 97%
2,4-Dichloro-6-iodoaniline, 97%
CAS: 697-90-5 Molecular Formula: C6H4Cl2IN Molecular Weight (g/mol): 287.909 MDL Number: MFCD00007661 InChI Key: YJRSMJTVXWBFJJ-UHFFFAOYSA-N Synonym: 2,4-dichloro-6-iodobenzenamine,2,4-dichloro-6-iodophenylamine,4,6-dichloro-2-iodophenylamine,2,4-dichloro-6-iodo-phenylamine,benzenamine,2,4-dichloro-6-iodo,benzenamine, 2,4-dichloro-6-iodo,1-amino-2,4-dichloro-6-iodobenzene PubChem CID: 7577525 IUPAC Name: 2,4-dichloro-6-iodoaniline SMILES: C1=C(C=C(C(=C1Cl)N)I)Cl