missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-6-iodoaniline, 97%

Catalog No. AAL1765914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1765914 25 g
AAL1765906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1765914 Supplier Thermo Scientific Chemicals Supplier No. L1765914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 697-90-5 | C6H4Cl2IN | 287.909 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 697-90-5
Molecular Formula C6H4Cl2IN
Molecular Weight (g/mol) 287.909
MDL Number MFCD00007661
InChI Key YJRSMJTVXWBFJJ-UHFFFAOYSA-N
Synonym 2,4-dichloro-6-iodobenzenamine, 2,4-dichloro-6-iodophenylamine, 4,6-dichloro-2-iodophenylamine, 2,4-dichloro-6-iodo-phenylamine, benzenamine,2,4-dichloro-6-iodo, benzenamine, 2,4-dichloro-6-iodo, 1-amino-2,4-dichloro-6-iodobenzene
PubChem CID 7577525
IUPAC Name 2,4-dichloro-6-iodoaniline
SMILES C1=C(C=C(C(=C1Cl)N)I)Cl

Specifications

Melting Point 79°C to 81°C
Quantity 25 g
UN Number UN2811
Beilstein 3246130
Sensitivity Light sensitive
Formula Weight 287.92
Percent Purity 97%
Chemical Name or Material 2,4-Dichloro-6-iodoaniline
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only