accessibility menu, dialog, popup

UserName

BrN

CAS RN 6952-59-6

IUPAC Name: 3-bromobenzonitrile
Synonyms: 3-bromobenzonitrile
Molecular Weight (g/mol): 182.02
Molecular Formula: C7H4BrN
InChi Key: STXAVEHFKAXGOX-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=C1)C#N
Boiling Point 
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (3)
  • (4)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)