accessibility menu, dialog, popup

UserName

CAS RN 6947-94-0

IUPAC Name: 2-methylfuran-3-carboxylate
Synonyms: 2-methylfuran-3-carboxylate
Molecular Weight (g/mol): 125.1
Molecular Formula: C6H5O3
InChi Key: CFGQZVOVFIZRMN-UHFFFAOYSA-M
SMILES: CC1=C(C=CO1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results