accessibility menu, dialog, popup

UserName

CAS RN 693-36-7

H3COOSOOCH3

CAS RN 693-36-7

IUPAC Name: octadecyl 3-{[3-(octadecyloxy)-3-oxopropyl]sulfanyl}propanoate
Synonyms: distearyl thiodipropionate
Molecular Weight (g/mol): 683.17
Molecular Formula: C42H82O4S
InChi Key: PWWSSIYVTQUJQQ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCOC(=O)CCSCCC(=O)OCCCCCCCCCCCCCCCCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results