accessibility menu, dialog, popup

UserName

CAS RN 691-24-7

IUPAC Name: tert-butyl[(tert-butylimino)methylidene]amine
Synonyms: tert-butyl[(tert-butylimino)methylidene]amine
Molecular Weight (g/mol): 154.26
Molecular Formula: C9H18N2
InChi Key: IDVWLLCLTVBSCS-UHFFFAOYSA-N
SMILES: CC(C)(C)N=C=NC(C)(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results