accessibility menu, dialog, popup

UserName

CAS RN 6898-84-6

IUPAC Name: sodium propylsulfanide
Synonyms: sodium propylsulfanide
Molecular Weight (g/mol): 98.14
Molecular Formula: C3H7NaS
InChi Key: XVSFHIIADLZQJP-UHFFFAOYSA-M
SMILES: [Na+].CCC[S-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results