accessibility menu, dialog, popup

UserName

CAS RN 68837-59-2

IUPAC Name: 4-bromo-2-methylbenzoic acid
Synonyms: 4-bromo-2-methylbenzoic acid
Molecular Weight (g/mol): 215.05
Molecular Formula: C8H7BrO2
InChi Key: RVCJOGNLYVNRDN-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=CC=C1C(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (1)
Quantity 
  • (2)
  • (1)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)