missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methylbenzoic Acid 98.0+%, TCI America™

Catalog No. B33375G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33375G 5 g
B333725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33375G Supplier TCI America Supplier No. B33375G

Chemical Identifiers

CAS 68837-59-2
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00040905
InChI Key RVCJOGNLYVNRDN-UHFFFAOYSA-N
Synonym 2-methyl-4-bromobenzoic acid, 4-bromo-o-toluic acid, 4-bromo-2-methyl-benzoic acid, benzoic acid, 4-bromo-2-methyl, synquest 2721-9-x6, 4-bromo-2-methylbenz, 4-bromo-2-methylbenzoicacid, pubchem4029, 5-bromo-2-carboxytoluene, acmc-1baz5
PubChem CID 99570
IUPAC Name 4-bromo-2-methylbenzoic acid
SMILES CC1=C(C=CC(=C1)Br)C(=O)O

Specifications

Melting Point 181°C
Color White-Yellow
Quantity 5 g
Formula Weight 215.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-methylbenzoic Acid