missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methylbenzoic acid, 97%

Catalog No. AC443400050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC443400050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC443400050 Supplier Thermo Scientific Chemicals Supplier No. 443400050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 68837-59-2 | C8H7BrO2 | 215.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68837-59-2
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
InChI Key RVCJOGNLYVNRDN-UHFFFAOYSA-N
Synonym 2-methyl-4-bromobenzoic acid, 4-bromo-o-toluic acid, 4-bromo-2-methyl-benzoic acid, benzoic acid, 4-bromo-2-methyl, synquest 2721-9-x6, 4-bromo-2-methylbenz, 4-bromo-2-methylbenzoicacid, pubchem4029, 5-bromo-2-carboxytoluene, acmc-1baz5
PubChem CID 99570
IUPAC Name 4-bromo-2-methylbenzoic acid
SMILES CC1=C(C=CC(=C1)Br)C(=O)O

Specifications

Melting Point 182.0°C to 184.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 215.05
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-methylbenzoic acid
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Toxic if swallowed. Causes serious eye irritation.
Precautionary Statement GHS P Statement Wear eye protection/face protection. Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. If eye irritation persists: Get medical advice/attention.
EINECSNumber 272-437-7

RUO – Research Use Only