accessibility menu, dialog, popup

UserName

SSSSSSSS

CAS RN 68550-20-9

IUPAC Name: 2-{2H,5H-[1,3]dithiolo[4,5-d][1,3]dithiol-2-ylidene}-2H,5H-[1,3]dithiolo[4,5-d][1,3]dithiole
Synonyms: 2-{5H-[1,3]dithiolo[4,5-d][1,3]dithiol-2-ylidene}-5H-[1,3]dithiolo[4,5-d][1,3]dithiole
Molecular Weight (g/mol): 356.6
Molecular Formula: C8H4S8
InChi Key: YAHVGMRXXZDSOK-UHFFFAOYSA-N
SMILES: C1SC2=C(S1)SC(S2)=C1SC2=C(SCS2)S1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results