accessibility menu, dialog, popup

UserName

CAS RN 68494-08-6

IUPAC Name: {[5-(benzoylsulfanyl)-2-sulfanylidene-2H-1,3-dithiol-4-yl]sulfanyl}(phenyl)methanone
Synonyms: {[5-(benzoylsulfanyl)-2-sulfanylidene-1,3-dithiol-4-yl]sulfanyl}(phenyl)methanone
Molecular Weight (g/mol): 406.57
Molecular Formula: C17H10O2S5
InChi Key: YQZPSSPUVUGQQS-UHFFFAOYSA-N
SMILES: O=C(SC1=C(SC(=O)C2=CC=CC=C2)SC(=S)S1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results