accessibility menu, dialog, popup

UserName

A(a)

CAS RN 68478-92-2

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 474.68
Molecular Formula: C8 H18 O Pt Si2
InChi Key: —
SMILES: —
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)