accessibility menu, dialog, popup

UserName

CAS RN 68399-81-5

HOOHHOHNHOSOHOO

CAS RN 68399-81-5

IUPAC Name: 3-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}-2-hydroxypropane-1-sulfonic acid
Synonyms: tapso
Molecular Weight (g/mol): 259.27
Molecular Formula: C7H17NO7S
InChi Key: RZQXOGQSPBYUKH-UHFFFAOYNA-N
SMILES: OCC(CO)(CO)NCC(O)CS(O)(=O)=O
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)
pH 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)