accessibility menu, dialog, popup

UserName

CAS RN 68157-60-8

IUPAC Name: 3-(2-chloropyridin-4-yl)-1-phenylurea
Synonyms: forchlorfenuron
Molecular Weight (g/mol): 247.68
Molecular Formula: C12H10ClN3O
InChi Key: GPXLRLUVLMHHIK-UHFFFAOYSA-N
SMILES: ClC1=CC(NC(=O)NC2=CC=CC=C2)=CC=N1
Percent Purity 
  • (3)
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results