missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Forchlorfenuron, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 165.0°C to 170.0°C |
| Solubility | Soluble in water, methanol, ethanol and acetone; Slightly soluble in chloroform |
| Solubility Information | Solubility in water: 0.039g/L. Other solubilities: soluble in methanol (119g/L), ethanol (149g/L),, acetone (127g/L);, slightly soluble in chloroform (2.7g/L) |
| Quantity | 25 g |
| Formula Weight | 247.68 |
| Percent Purity | ≥97.5% (HPLC) |
| Chemical Name or Material | Forchlorfenuron |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement Suspected of causing cancer. Toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Obtain special instructions before use. Use personal protective equipment as required. IF exposed or concerned: Get medical advice/attention. Avoid release to the environment. |
RUO – Research Use Only