missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Chloro-4-pyridyl)-3-phenylurea 98.0+%, TCI America™

Catalog No. C09265G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C09265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C09265G Supplier TCI America Supplier No. C09265G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A plant growth regulator from the cytokinin class, suitable for laboratory research applications.

Specifications

CAS 68157-60-8
Melting Point 172°C
Color White
Molecular Formula C12H10ClN3O
Quantity 5 g
MDL Number MFCD00059898
UN Number 3077
Synonym forchlorfenuron, 1-2-chloropyridin-4-yl-3-phenylurea, 1-2-chloro-4-pyridyl-3-phenylurea, cppu, 4pu30 cpd, n-2-chloro-4-pyridyl-n'-phenylurea, urea, n-2-chloro-4-pyridinyl-n'-phenyl, unii-k62ip7463j, 4-cppu, ccpu
InChI Key GPXLRLUVLMHHIK-UHFFFAOYSA-N
SMILES ClC1=CC(NC(=O)NC2=CC=CC=C2)=CC=N1
IUPAC Name 3-(2-chloropyridin-4-yl)-1-phenylurea
Molecular Weight (g/mol) 247.68
PubChem CID 93379
ChEBI CHEBI:81861
Formula Weight 247.68
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2-Chloro-4-pyridyl)-3-phenylurea
Show More Show Less
RTECSNumber YS7182500
TSCA No
Recommended Storage Refrigerator