accessibility menu, dialog, popup

UserName

CoNNNNNN(S)P(R)(R)(S)O(R)O(R)ONHNOOOOOOOHOOOH3CH3CCH3H3CCH3CH3NH2NH2NH2H2NH2NH2NH3CH3COHH3CH3CCH3HHHHHHHH

CAS RN 68-19-9

IUPAC Name: (10S,12R,13R,24S,35R,41R)-30,35,40-tris(2-carbamoylethyl)-24,36,41-tris(carbamoylmethyl)-1-cyano-46-hydroxy-12-(hydroxymethyl)-5,6,17,23,28,31,31,36,38,41,42-undecamethyl-15,20-dioxo-11,14,16-trioxa-2λ⁵,9,19,26λ⁵,43,44λ⁵,45λ⁵-heptaaza-15λ⁵-phospha-1-cobaltadodecacyclo[27.14.1.1¹,³⁴.1²,⁹.1¹⁰,¹³.0¹,²⁶.0³,⁸.0²³,²⁷.0²⁵,⁴².0³²,⁴⁴.0³⁹,⁴³.0³⁷,⁴⁵]heptatetraconta-2(47),3,5,7,26,28,32(44),33,37(45),38-decaene-1,2,26,44,45-pentakis(ylium)-15-olate
Synonyms: (10S,12R,13R,24S,35R,41R)-30,35,40-tris(2-carbamoylethyl)-24,36,41-tris(carbamoylmethyl)-1-cyano-46-hydroxy-12-(hydroxymethyl)-5,6,17,23,28,31,31,36,38,41,42-undecamethyl-15,20-dioxo-11,14,16-trioxa-2λ⁵,9,19,26λ⁵,43,44λ⁵,45λ⁵-heptaaza-15λ⁵-phospha-1-cobaltadodecacyclo[27.14.1.1¹,³⁴.1²,⁹.1¹⁰,¹³.0¹,²⁶.0³,⁸.0²³,²⁷.0²⁵,⁴².0³²,⁴⁴.0³⁹,⁴³.0³⁷,⁴⁵]heptatetraconta-2(47),3,5,7,26,28,32(44),33,37(45),38-decaene-1,2,26,44,45-pentakis(ylium)-15-olate
Molecular Weight (g/mol): 1355.39
Molecular Formula: C63H88CoN14O14P
InChi Key: YUWVGNQGDAPKCX-BVWPOUIRNA-L
SMILES: [C@@H]1(C2=CC3=[N+]4C(C(C3(C)C)CCC(N)=O)=C(C3=[N+]5C6C7(N8C(C([C@@]7(C)CC(N)=O)CCC(N)=O)=C(C(=[N+]2[Co+]458([N+]2=CN(C4=CC(=C(C)C=C24)C)[C@@]2([H])O[C@@H]([C@]([H])(OP([O-])(=O)OC(C)CNC(=O)CCC3(C)[C@@H]6CC(N)=O)C2O)CO)C#N)C1(C)CC(N)=O)C)C)C)CCC(N)=O
Molecular Weight (g/mol) 
  • (13)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (4)
  • (1)
  • (4)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
Physical Form 
  • (5)
  • (4)
Grade 
  • (1)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (11)
  • (5)
  • (1)
  • (1)
  • (4)
  • (2)
  • (7)
  • (13)
  • (1)
  • (2)
  • (3)
  • (4)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (5)
  • (4)
  • (1)
  • (5)