accessibility menu, dialog, popup

UserName

NH2H2NH2O

CAS RN 6780-13-8

IUPAC Name: ethane-1,2-diamine hydrate
Synonyms: ethylenediamine hydrate
Molecular Weight (g/mol): 78.12
Molecular Formula: C2H10N2O
InChi Key: XZUAPPXGIFNDRA-UHFFFAOYSA-N
SMILES: O.NCCN
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results