accessibility menu, dialog, popup

UserName

H3CNOClClClONN

CAS RN 67747-09-5

IUPAC Name: N-propyl-N-[2-(2,4,6-trichlorophenoxy)ethyl]-1H-imidazole-1-carboxamide
Synonyms: prochloraz mirage (pesticide) epa pesticide chemical code 128851 prelude sporgon prochloraz-mn caswell no. 704E sportak PF mirage sportak
Molecular Weight (g/mol): 376.66
Molecular Formula: C15H16Cl3N3O2
InChi Key: TVLSRXXIMLFWEO-UHFFFAOYSA-N
SMILES: CCCN(CCOC1=C(Cl)C=C(Cl)C=C1Cl)C(=O)N1C=CN=C1
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results