missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prochloraz 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 50°C |
| Quantity | 5 g |
| UN Number | 3077 |
| Formula Weight | 376.66 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Prochloraz |
Safety and Handling
| RTECSNumber | NI4000400 |
| TSCA | No |
| Recommended Storage | Refrigerator |