missing translation for 'onlineSavingsMsg'
Learn More

Prochloraz, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111019000
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019000 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019000 Supplier Merck Emd Millipore Supplier No. 6494750MG
Only null left

Prochloraz is a fungicide possessing imidazole moiety, which can show anti-androgenic activity in blocking androgens like testosterone from mediating their biological effects in the body. It is involved in the inhibition of cytochrome P450-dependent, 14 α-demethylase activity required in the conversion of lanosterol to ergosterol, a key constituent of fungal cell membranes.

Specifications

Packaging Bottomless Glass Bottle with Fused Cone
CAS 67747-09-5
Product Line TraceCERT
Physical Form Neat
Linear Formula C15H16Cl3N3O2
MDL Number MFCD00078735
Quantity 50 mg
Formula Weight 376.67
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC