accessibility menu, dialog, popup

UserName

BrOBr

CAS RN 67713-23-9

IUPAC Name: 2,6-dibromobenzaldehyde
Synonyms: 2,6-dibromobenzaldehyde
Molecular Weight (g/mol): 263.92
Molecular Formula: C7H4Br2O
InChi Key: YDYNSAUGVGAOLO-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(Br)=C1C=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results