missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromobenzaldehyde 98.0+%, TCI America™

Catalog No. D45525G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45525G 5 g
D45521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45525G Supplier TCI America Supplier No. D45525G

Chemical Identifiers

CAS 67713-23-9
Molecular Formula C7H4Br2O
Molecular Weight (g/mol) 263.92
MDL Number MFCD11040410
InChI Key YDYNSAUGVGAOLO-UHFFFAOYSA-N
PubChem CID 12648641
IUPAC Name 2,6-dibromobenzaldehyde
SMILES BrC1=CC=CC(Br)=C1C=O

Specifications

Melting Point 92°C
Color Yellow
Quantity 5 g
Formula Weight 263.92
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dibromobenzaldehyde