accessibility menu, dialog, popup

UserName

CAS RN 677-43-0

IUPAC Name: (dimethylamino)phosphonoyl dichloride
Synonyms: dimethylamidophosphoric dichloride
Molecular Weight (g/mol): 161.95
Molecular Formula: C2H6Cl2NOP
InChi Key: YNHXBEVSSILHPI-UHFFFAOYSA-N
SMILES: CN(C)P(Cl)(Cl)=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)