accessibility menu, dialog, popup

UserName

CAS RN 6757-06-8

IUPAC Name: disodium [(2R,3S,4R,5R)-5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
Synonyms: disodium 5'-cytidylate
Molecular Weight (g/mol): 367.16
Molecular Formula: C9H12N3Na2O8P
InChi Key: INTPYBRGLGSMRA-IJRYJGAHNA-L
SMILES: [Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]1O
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (5)
Quantity 
  • (2)
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (5)
  • (2)
  • (1)
  • (2)
  • (2)