accessibility menu, dialog, popup

UserName

CAS RN 675-14-9

IUPAC Name: trifluoro-1,3,5-triazine
Synonyms: cyanuric fluoride
Molecular Weight (g/mol): 135.05
Molecular Formula: C3F3N3
InChi Key: VMKJWLXVLHBJNK-UHFFFAOYSA-N
SMILES: FC1=NC(F)=NC(F)=N1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results