accessibility menu, dialog, popup

UserName

CAS RN 674783-97-2

IUPAC Name: 10-methyl-9-(2,4,6-trimethylphenyl)acridin-10-ium perchlorate
Synonyms: 10-methyl-9-(2,4,6-trimethylphenyl)acridin-10-ium perchlorate
Molecular Weight (g/mol): 411.88
Molecular Formula: C23H22ClNO4
InChi Key: LFMBERYWDLWXNO-UHFFFAOYSA-M
SMILES: [O-][Cl](=O)(=O)=O.CC1=CC(C)=C(C(C)=C1)C1=C2C=CC=CC2=[N+](C)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results