accessibility menu, dialog, popup

UserName

BrSFFFFF

CAS RN 672-30-0

IUPAC Name: 1-bromo-3-(pentafluoro-λ⁶-sulfanyl)benzene
Synonyms: 1-bromo-3-(pentafluoro-λ⁶-sulfanyl)benzene
Molecular Weight (g/mol): 283.05
Molecular Formula: C6H4BrF5S
InChi Key: QRPMKEUTGAXKSD-UHFFFAOYSA-N
SMILES: FS(F)(F)(F)(F)C1=CC(Br)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results