missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromophenylsulfur Pentafluoride 95.0+%, TCI America™
Specifications
Specifications
| Boiling Point | 82°C |
| Quantity | 1 g |
| Formula Weight | 283.05 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 3-Bromophenylsulfur Pentafluoride |
Safety and Handling
| TSCA | No |