accessibility menu, dialog, popup

UserName

CAS RN 672-13-9

IUPAC Name: 2-hydroxy-5-methoxybenzaldehyde
Synonyms: 2-hydroxy-5-methoxybenzaldehyde
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: FZHSPPYCNDYIKD-UHFFFAOYSA-N
SMILES: COC1=CC=C(O)C(C=O)=C1
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (2)
  • (7)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
Boiling Point 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (7)
  • (6)
  • (3)
  • (2)
  • (7)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (4)
  • (5)