accessibility menu, dialog, popup

UserName

CAS RN 67-51-6

IUPAC Name: 3,5-dimethyl-1H-pyrazole
Synonyms: 1H-pyrazole, 3,5-dimethyl-
Molecular Weight (g/mol): 96.13
Molecular Formula: C5H8N2
InChi Key: SDXAWLJRERMRKF-UHFFFAOYSA-N
SMILES: CC1=CC(C)=NN1
Boiling Point 
  • (6)
  • (2)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (6)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)