accessibility menu, dialog, popup

UserName

CAS RN 66946-49-4

IUPAC Name: 2-{2H,5H,6H,7H-[1,3]dithiolo[4,5-b][1,4]dithiepin-2-ylidene}-2H,5H,6H,7H-[1,3]dithiolo[4,5-b][1,4]dithiepine
Synonyms: 2-{5H,6H,7H-[1,3]dithiolo[4,5-b][1,4]dithiepin-2-ylidene}-5H,6H,7H-[1,3]dithiolo[4,5-b][1,4]dithiepine
Molecular Weight (g/mol): 412.71
Molecular Formula: C12H12S8
InChi Key: LSZPCLOYNFOXST-UHFFFAOYSA-N
SMILES: C1CSC2=C(SC(S2)=C2SC3=C(S2)SCCCS3)SC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results