accessibility menu, dialog, popup

UserName

CAS RN 66-71-7

IUPAC Name: 1,10-phenanthroline
Synonyms: phen 2-phenanthroline 1,10-fenanthrolin β-phenanthroline orthophenanthroline O-phenanthroline o-phenanthroline O-phe zinc salt 1,10-phenanthroline
Molecular Weight (g/mol): 180.21
Molecular Formula: C12H8N2
InChi Key: DGEZNRSVGBDHLK-UHFFFAOYSA-N
SMILES: C1=CN=C2C(C=CC3=CC=CN=C23)=C1
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (2)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (7)
  • (3)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (4)
  • (3)