accessibility menu, dialog, popup

UserName

CAS RN 65934-74-9

IUPAC Name: 4-methyl-3-(trifluoromethyl)aniline
Synonyms: 4-methyl-3-(trifluoromethyl)aniline
Molecular Weight (g/mol): 175.15
Molecular Formula: C8H8F3N
InChi Key: JBCDCYFEJQHTTA-UHFFFAOYSA-N
SMILES: CC1=CC=C(N)C=C1C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results