accessibility menu, dialog, popup

UserName

CAS RN 65184-12-5

HOHNOH

CAS RN 65184-12-5

IUPAC Name: 2-[(hydroxymethyl)amino]ethan-1-ol
Synonyms: 2-[(hydroxymethyl)amino]ethanol
Molecular Weight (g/mol): 91.11
Molecular Formula: C3H9NO2
InChi Key: NWPCFCBFUXXJIE-UHFFFAOYSA-N
SMILES: OCCNCO
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results