accessibility menu, dialog, popup

UserName

CAS RN 65141-46-0

IUPAC Name: 2-[(pyridin-3-yl)formamido]ethyl nitrate
Synonyms: 2-nicotinamidoethyl nitrate
Molecular Weight (g/mol): 211.18
Molecular Formula: C8H9N3O4
InChi Key: LBHIOVVIQHSOQN-UHFFFAOYSA-N
SMILES: [O-][N+](=O)OCCNC(=O)C1=CC=CN=C1
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)