accessibility menu, dialog, popup

UserName

CAS RN 650-51-1

IUPAC Name: sodium trichloroacetate
Synonyms: sodium trichloroacetate
Molecular Weight (g/mol): 185.36
Molecular Formula: C2Cl3NaO2
InChi Key: SAQSTQBVENFSKT-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)C(Cl)(Cl)Cl
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)