accessibility menu, dialog, popup

UserName

CAS RN 65-71-4

IUPAC Name: 5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: thymine Thy
Molecular Weight (g/mol): 126.12
Molecular Formula: C5H6N2O2
InChi Key: RWQNBRDOKXIBIV-UHFFFAOYSA-N
SMILES: CC1=CNC(=O)NC1=O
Molecular Weight (g/mol) 
  • (4)
  • (8)
Percent Purity 
  • (3)
  • (2)
  • (3)
  • (4)
Quantity 
  • (1)
  • (3)
  • (4)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (3)
  • (7)
  • (4)
  • (8)
  • (3)
  • (2)
  • (3)
  • (4)
  • (1)
  • (3)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (4)